2',6'-Dimethoxyacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2040-04-2
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00008729
:180.2 g/mol
Boiling Pt:135…136 °C (2 mmHg)
Melting Pt:68…71 °C
Storage Temperature:Ambient
EINECS:218-034-1
Related Information
Frequently Bought Together