3'-Nitroacetophenone 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: m-Nitroacetophenone
- CAS Number:121-89-1
- Formula:C₈H₇NO₃
- MDL Number:MFCD00007259
- Molecular Weight:165.15 g/mol
- Boiling Pt:202 °C (1013 hPa)
- Melting Pt:76…79 °C
- Density:1,433 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-504-3
2 Options Available Below
Product Details & Documents
Synonyms: m-Nitroacetophenone
- CAS Number:121-89-1
- Formula:C₈H₇NO₃
- MDL Number:MFCD00007259
- Molecular Weight:165.15 g/mol
- Boiling Pt:202 °C (1013 hPa)
- Melting Pt:76…79 °C
- Density:1,433 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-504-3
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to yellow-green to yellow-brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:76 to 80 °C
- GC:≥97.5 %



