Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
2'-Nitroacetophenone 95%

2'-Nitroacetophenone 95%

Irritant
2'-Nitroacetophenone 95%
Certificates
2'-Nitroacetophenone 95%

2 Options Available Below

Product Details & Documents
Synonyms:  o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone
  • CAS Number:577-59-3
  • Formula:C₈H₇NO₃
  • MDL Number:MFCD00007144
  • Molecular Weight:165.15 g/mol
  • Boiling Pt:159 °C (16 mmHg)
  • Melting Pt:28…30 °C
  • Density:1,24 g/cm³ (20 °C)
  • Flash Pt:>112 °C
  • Storage Temperature:Refrigerator
  • EINECS:209-414-8

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Orange to brown Low melting solid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥94 %
  • Stabilizer:
    0.05 % BHT
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    2,5 g
    Log in for availability
    Log in for availability
    1 * 2,5 g
    1 * 2,5 g - £42.00
    £42.00
    1 * 2,5 g
       £42.00

  • Glass bottle for solids
    20 g
    Log in for availability
    Log in for availability
    1 * 20 g
    1 * 20 g - £148.00
    £148.00
    1 * 20 g
       £148.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.