4-Chloro-m-cresol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 4-Chloro-3-methylphenol, 2-Chloro-5-hydroxytoluene, 1-Chloro-4-hydroxy-2-methylbenzene, p-Chloro m-cresol
CAS Number:59-50-7
:
Formula:C₇H₇ClO
MDL Number:MFCD00002323
Merck Index:13,02152
Boiling Pt:235 °C (1013 hPa)
Melting Pt:63…65 °C
Density:0,9 g/cm³ (20 °C)
Flash Pt:118 °C
EINECS:200-431-6
UN:3437
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:142.58 g/mol
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:3.9 g/l (20 °C)
- Melting Point:63 - 65 °C
- Molar Mass:142.58 g/mol
- Bulk Density:800 kg/m3
- Boiling Point:235 - 239 °C (1013 hPa)
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:118 °C
- Density:1.37 g/cm3 (20 °C)
- pH:6.5 (1 g/l, H2O, 20 °C)
- Ignition Point:590 °C
Specification Test Results
Related Information
Frequently Bought Together