5-Chloro-o-cresol
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: 1-Chloro-3-hydroxy-4-methylbenzene, 5-Chloro-2-methylphenol
CAS Number:5306-98-9
:
Formula:C₇H₇ClO
MDL Number:MFCD00060672
Boiling Pt:225
Molecular Weight:142.58 g/mol
Melting Pt:75…76 °C
EINECS:226-160-3
UN:3437
ADR:6.1 II
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together