


4-Chloro-3-(trifluoromethyl)phenyl isocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:327-78-6
:
Formula:C₈H₃ClF₃NO
MDL Number:MFCD00013874
:221.57 g/mol
Boiling Pt:86…90 °C (14 mmHg)
Melting Pt:34…36 °C
Flash Pt:99 °C (210 °F)
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together