

2-Chloro-5-(trifluoromethyl)aniline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:121-50-6
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD00007673
Boiling Pt:82…83 °C (9 mmHg)
Melting Pt:8…10 °C
Density:1,424
Flash Pt:75 °C (167 °F)
Storage Temperature:Ambient
EINECS:204-475-7
ADR:6.1,III
:195.57 g/mol
UN:2810
:GHS06
:GHS07
Related Information
Frequently Bought Together