(3,4-Dichlorophenyl)acetonitrile ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3218-49-3
:
Formula:C₈H₅Cl₂N
MDL Number:MFCD00001909
:186.04 g/mol
Boiling Pt:169…171 °C (12 mmHg)
Melting Pt:38…42 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:221-743-9
UN:3439
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together