

2,6-Dibromo-4-isopropylaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10546-65-3
:
Formula:C₉H₁₁Br₂N
MDL Number:MFCD00015475
Boiling Pt:316 °C (1013 hPa)
Melting Pt:45…47 °C
:293 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together