
ω-Bromo-2,4-dimethoxyacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60965-26-6
:
Formula:C₁₀H₁₁BrO₃
MDL Number:MFCD00000197
:259.1 g/mol
Boiling Pt:315 °C (1013 hPa)
Melting Pt:102…105 °C
EINECS:262-542-6
UN:3261
ADR:8,III
:GHS05
Related Information
Frequently Bought Together