1,2-Benzenedithiol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17534-15-5
:
Formula:C₆H₆S₂
MDL Number:MFCD00004835
:142.25 g/mol
Boiling Pt:239 °C
Melting Pt:21…25 °C
Density:1,236
Flash Pt:103 °C (217 °F)
EINECS:241-530-4
ADR:9,III
:GHS05
:GHS07
Related Information
Frequently Bought Together