Phloroglucinol, Sigma-Aldrich®
: 1,3,5-Trihydroxybenzene
:108-73-6
:
:C₆H₆O₃
:MFCD00002286
:400
:215…220 °C
:1.46 g/cm³ (20 °C)
:126.11 g/mol
:12,07482
:M-Clarity™ quality level = MQ100
:203-611-2
:GHS07
:Room Temperature
- Environmentally Preferable:
- Appearance (colour):White to faint yellow to faint beige
- Appearance (form):Powder or crystals
- Purity (HPLC area %):Report result
- Solubility (colour):Colorless to light brown-yellow
- Solubility (turbidity):Clear to hazy
- Solubility (method):50 mg/ml ethanol
- Loss on drying:Report result
- Carbon content:Report result
- Infrared spectrum:Conforms to structure
Frequently Bought Together