
1,2,4-Trihydroxybenzene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:533-73-3
:
Formula:C₆H₆O₃
MDL Number:MFCD00002198
:126.11 g/mol
Merck Index:14,1073
Boiling Pt:362 °C (1013 hPa)
Melting Pt:∼ 140 °C (subl.)
Density:1,45 g/cm³ (20 °C)
EINECS:208-575-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together