


Phthalic anhydride 99.0-100.2% ACS
Catalog # 41771.36
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
CAS Number:85-44-9
:
Formula:C₈H₄O₃
MDL Number:MFCD00005918
:148.12 g/mol
Merck Index:14,7372
Boiling Pt:295 °C (subl.)
Melting Pt:131…133 °C
Density:1,53
Flash Pt:152 °C (305 °F)
Storage Temperature:Ambient
EINECS:201-607-5
UN:2214
ADR:8,III
:GHS08
:GHS05
:GHS07