Order Entry
Northern Ireland
ContactUsLinkComponent
Phthalic anhydride ACS
Health
Corrosive
Irritant
Phthalic anhydride ACS
Certificates
undefined
Health
Corrosive
Irritant
Phthalic anhydride ACS
Certificates

Specification Test Results

  • Appearance:
    White Flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    129 to 132 °C
  • Residue after ignition:
    ≤0.01 %
  • Titration by coupling:
    99.0 to 100.2 %
  • Chloride (Cl):
    ≤0.002 %
  • Sulfate (SO4):
    ≤0.003 %
  • Iron (Fe):
    ≤5 ppm
  • Heavy metals (as Pb):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together