
2-Phenoxyethanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122-99-6
:
Formula:C₆H₅OCH₂CH₂OH
MDL Number:MFCD00002857
:138.17 g/mol
Merck Index:12,07410
Boiling Pt:237 °C (760 mmHg)
Melting Pt:11…13 °C
Density:1,1 g/cm³
Flash Pt:130 °C
EINECS:204-589-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5360 to 1.5400 (20°C, 589 nm)
- GC:≥98.5 %
- pH:9.5 to 11.0 (5% at 25°C)
- Titration with HClO4:≥99 %
- Iron (Fe):≤0.001 %
Specification Test Results
Frequently Bought Together