Order Entry
Northern Ireland
ContactUsLinkComponent
p-Phenylenediamine for synthesis, Sigma-Aldrich®
p-Phenylenediamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
p-Phenylenediamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    47 g/l (25 °C)
  • Melting Point:
    139 - 141 °C
  • Molar Mass:
    108.14 g/mol
  • Bulk Density:
    600 kg/m3
  • Boiling Point:
    267 °C (1013 hPa)
  • Vapor Pressure:
    0.01 hPa (20 °C)
  • Flash Point:
    156 °C
  • Density:
    1.135 g/cm3 (20 °C)
  • pH:
    9 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    567 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together