


m-Phenylenediamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-45-2
:
Formula:C₆H₈N₂
MDL Number:MFCD00007799
:108.14 g/mol
Merck Index:12,07437
Boiling Pt:282 °C (1013 hPa)
Melting Pt:63…64 °C
Density:1,14 g/cm³ (20 °C)
EINECS:203-584-7
UN:1673
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:187 °C
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:350 g/l (25 °C)
- Melting Point:63 - 64 °C
- Molar Mass:108.14 g/mol
- Bulk Density:600 kg/m3
- Boiling Point:283 - 284 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (100 °C)
- Flash Point:187 °C
- Density:1.14 g/cm3 (20 °C)
- pH:8 (100 g/l, H2O, 20 °C)
- Ignition Point:560 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Frequently Bought Together