Order Entry
Northern Ireland
ContactUsLinkComponent
m-Phenylenediamine 99+%
Toxic
Health
Environment
m-Phenylenediamine 99+%
Certificates
undefined
Toxic
Health
Environment
m-Phenylenediamine 99+%

Specification Test Results

  • Appearance:
    White to tan to grey Flakes or pellets (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    62 to 65 °C
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.001 %

Specification Test Results

Related Information

Frequently Bought Together