1,4-Phenylene diisothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4044-65-9
:
Formula:C₈H₄N₂S₂
MDL Number:MFCD00004811
:192.27 g/mol
Merck Index:14,1308
Boiling Pt:319 °C (1013 hPa)
Melting Pt:128…133 °C
EINECS:223-741-3
UN:2923
ADR:8,III
:GHS06
:GHS08
:GHS05
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together