1,4-Phenylene diisothiocyanate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4044-65-9
:
Formula:C₈H₄N₂S₂
MDL Number:MFCD00004811
:192.27 g/mol
Merck Index:13,01304
Boiling Pt:319 °C (1013 hPa)
Melting Pt:129…133 °C
EINECS:223-741-3
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Off-white to yellow to beige to light grey Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:129 to 133 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together