
Phenyl isothiocyanate for synthesis, Sigma-Aldrich®
: Merck
:103-72-0
:
:C₇H₅NS
:MFCD00004798
:135.19 g/mol
:12,07450
:221 °C (1013 hPa)
:–21 °C
:1,13 g/cm³ (20 °C)
:Refrigerator
:203-138-1
:2206
:6.1,II
:M-Clarity™ quality level = MQ200
:87 °C
:GHS06
- Solubility:(20 °C) insoluble,(decomposition)
- Melting Point:-21 °C
- Molar Mass:135.18 g/mol
- Boiling Point:221 °C (1013 hPa)
- Vapor Pressure:10 hPa (20 °C)
- Flash Point:87 °C
- Refractive Index:1.6497 (20 °C, 589 nm)
- Density:1.13 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Synonyms:Isothiocyanic acid phenyl ester, PTC
- Hill Formula:C7H5NS
- Chemical Formula:C6H5NCS
Frequently Bought Together