


2-Methoxyphenyl isocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:700-87-8
:
Formula:C₈H₇NO₂
MDL Number:MFCD00002004
:149.15 g/mol
Boiling Pt:94…96 °C (10 mmHg)
Density:1,122
Flash Pt:>110 °C (230 °F)
EINECS:211-853-5
UN:2206
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together