
Ethyl-4-iodobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51934-41-9
:
Formula:C₉H₉IO₂
MDL Number:MFCD00017344
:276.07 g/mol
Boiling Pt:283 °C
Density:1,660
Flash Pt:>110 °C (230 °F)
EINECS:429-550-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together