Order Entry
Northern Ireland
ContactUsLinkComponent
1,4-Dimethoxy-2-fluorobenzene ≥97%
Irritant
1,4-Dimethoxy-2-fluorobenzene ≥97%
Catalog # L13505.06
CAS Number:  
Irritant
1,4-Dimethoxy-2-fluorobenzene ≥97%
Catalog # L13505.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:82830-49-7
 :
Formula:C₈H₉FO₂
MDL Number:MFCD00042487
 :156.16 g/mol
Boiling Pt:119…121 °C (40 mmHg)
Melting Pt:23…26 °C
 :GHS07
Storage Temperature:Ambient