Potassium hydrogen phthalate
Catalog # PROONIST-2185
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:60 g
- Seq:3377
Specifications
About this item
:
:204.22 g/mol
Merck Index:13,07694
Melting Pt:295…300 °C
Density:1,6362 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-889-4
CAS Number:877-24-7
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070