Potassium hydrogen phthalate 99.95-100.05% (dried basis), ULTREX®, Ultrapure, J.T.Baker®
Catalog # 4889-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:99.95 - 100.05%
- Assay (1-KOCOC₆H₄-2-COOH) (dried basis):99.95 - 100.05 %
- Identification (by IR):Passes Test
- pH of 0.05M Solution at 25°C:4.00 - 4.02
Specification Test Results
About this item
:
:204.22 g/mol
Merck Index:13,07694
Melting Pt:295…300 °C
Density:1,64 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-889-4
CAS Number:877-24-7
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070