Potassium hydrogen phthalate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:877-24-7
:
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070
:204.22 g/mol
Merck Index:14,7612
Melting Pt:297 °C (dec.)
Density:1,636
Storage Temperature:Ambient
EINECS:212-889-4
Related Information
Frequently Bought Together