Ethyl-4-nitrobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-77-4
:
Formula:C₉H₉NO₄
MDL Number:MFCD00007351
Merck Index:14,3832
Melting Pt:55…58 °C
:195.17 g/mol
Storage Temperature:Ambient
EINECS:202-786-2
Related Information
Frequently Bought Together