
Methyl-3-aminobenzoate 98%
:4518-10-9
:
:C₈H₉NO₂
:MFCD00017102
:151.16 g/mol
:120 °C (0,7 mmHg)
:52…55 °C
:1,232 g/cm³ (20 °C)
:224-842-5
:2811
:GHS07
- Appearance:Light beige to brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:49 to 54 °C
- Titration with HClO4:≥97.5 %
- Enantiomeric excess:≥97.5 % (GC)
Frequently Bought Together