Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 3-aminobenzoate
Ethyl-3-aminobenzoate 98%
Ethyl-3-aminobenzoate 98%
:
Thermo Fisher Scientific
Ethyl-3-aminobenzoate 98%
:
Thermo Fisher Scientific
:
582-33-2
:
:
C₉H₁₁NO₂
:
MFCD00007794
:
165.19 g/mol
:
294 °C
:
27…30 °C
:
1,106
:
>110 °C (230 °F)
:
209-482-9
:
GHS07
:
Ambient
msds_ALFAA14448
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Toluene ≥98%, TECHNICAL
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
1-Propanol, LiChrosolv® for HPLC, Supelco®
: Merck
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Methanol, SupraSolv® for gas chromatography, Supelco®
: Merck
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Acetylacetone, EMSURE® for analysis, Supelco®
: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals