Order Entry
Northern Ireland
ContactUsLinkComponent
Ethyl-4-(dimethylamino)benzoate 99+%
Ethyl-4-(dimethylamino)benzoate 99+%
Certificates
undefined
Ethyl-4-(dimethylamino)benzoate 99+%

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    62 to 65 °C
  • GC:
    ≥99.0 %
  • pH:
    5.0 to 7.0 (2 % solution at 25.0 °C)
  • Sensitivity:
    Passes test

Specification Test Results

Related Information

Frequently Bought Together