

2-Anisoyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21615-34-9
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00000664
:170.6 g/mol
Boiling Pt:128…129 °C (8 mmHg)
Density:1,146
Flash Pt:84 °C (184 °F)
Storage Temperature:Ambient
EINECS:244-477-5
UN:1729
ADR:8,II
:GHS05
:GHS07
Related Information
Frequently Bought Together