
2-Chlorobenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2905-23-9
:
Formula:C₆H₄Cl₂O₂S
MDL Number:MFCD00051520
:211.07 g/mol
Boiling Pt:150…152 °C (12 mmHg)
Melting Pt:26…28 °C
Density:1,548
Flash Pt:140 °C (284 °F)
Storage Temperature:Ambient
EINECS:220-807-3
UN:3265
ADR:8,II
:GHS05
Related Information
Frequently Bought Together