£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
2,3,4-Trichlorophenyl isothiocyanate
2,3,4-Trichlorophenyl isothiocyanate ≥97%
2,3,4-Trichlorophenyl isothiocyanate ≥97%
2,3,4-Trichlorophenyl isothiocyanate ≥97%
L12119.06
:
Thermo Fisher Scientific
:
2,3,4-Trichlorophenyl isothiocyanate ≥97%
L12119.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
238.52 g/mol
:
305 °C (1013 hPa)
:
68…71 °C
:
Ambient
:
2923
:
8,III
:
GHS06
:
GHS05
:
127142-69-2
:
C₇H₂Cl₃NS
:
MFCD00041049