Order Entry
Northern Ireland
ContactUsLinkComponent
4-Chlorophenyl isocyanate 98%
Toxic
Health
Corrosive
Environment
4-Chlorophenyl isocyanate 98%
Catalog # ACRO166560250
CAS Number:  
undefined
Toxic
Health
Corrosive
Environment
4-Chlorophenyl isocyanate 98%
Catalog # ACRO166560250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to yellow Crystalline solid (May develop some turbidity or precipitate)
  • Infrared spectrum:
    Conforms
  • Melting point:
    29 to 31 °C
  • GC:
    ≥97.5 %
  • Heavy metals (as Pb):
    ≤10 ppm

Specification Test Results

About this item

 :
 :153.57 g/mol
Boiling Pt:203…204 °C (760 mmHg)
Melting Pt:29…31 °C
Density:1,26 g/cm³
Flash Pt:90 °C
Storage Temperature:Ambient
EINECS:203-176-9
UN:2811
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS05
 :GHS09
CAS Number:104-12-1
Formula:C₇H₄ClNO
MDL Number:MFCD00002024