


2,6-Dichlorophenyl isocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39920-37-1
:
Formula:C₇H₃Cl₂NO
MDL Number:MFCD00002003
:188.01 g/mol
Boiling Pt:101 °C (5 mmHg)
Melting Pt:42…46 °C
Flash Pt:76 °C (168 °F)
EINECS:254-699-4
UN:2250
ADR:6.1,II
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together