


4-Bromo-1-butene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5162-44-7
:
Formula:C₄H₇Br
MDL Number:MFCD00000258
:135 g/mol
Boiling Pt:98…100 °C
Density:1,330
Flash Pt:1 °C (34 °F)
EINECS:225-937-4
UN:1993
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together