Ethyl-3-chlorobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1128-76-3
:
Formula:C₉H₉ClO₂
MDL Number:MFCD00013634
:184.62 g/mol
Boiling Pt:118…120 °C (13 mmHg)
Density:1,186
Storage Temperature:Ambient
EINECS:214-441-3
Related Information
Frequently Bought Together