

2-Chlorobenzoyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:609-65-4
:
Formula:C₇H₄Cl₂O
MDL Number:MFCD00000660
:175.01 g/mol
Boiling Pt:237…239 °C
Melting Pt:–4 °C
Density:1,379
Flash Pt:125 °C (257 °F)
Storage Temperature:Ambient
EINECS:210-194-0
UN:3265
ADR:8,II
:GHS05
:GHS07
Related Information
Frequently Bought Together