Terephthalaldehyde for synthesis, Sigma-Aldrich®
Catalog # 8.08617.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:3 g/l (50 °C)
- Melting Point:112 - 115 °C
- Molar Mass:134.13 g/mol
- Boiling Point:245 - 248 °C (1013 hPa)
Specification Test Results
About this item
:
:134.13 g/mol
Boiling Pt:245 °C (1013 hPa)
Density:1,381 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:210-784-8
CAS Number:623-27-8
Formula:C₈H₆O₂
MDL Number:MFCD00006949