4-Bromothioanisole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Bromo-4-(methylthio)benzene, 4-Bromophenyl methyl sulphide
CAS Number:104-95-0
:
Formula:C₇H₇BrS
MDL Number:MFCD00000102
Boiling Pt:242
Molecular Weight:203.1 g/mol
Melting Pt:35…40 °C
Flash Pt:>110 °C
EINECS:203-255-8
UN:3335
ADR:9 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals and/or chunks or low melting crystalline mass
- Infrared spectrum:Conforms
- Melting point:35 to 40 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together