ContactUsLinkComponent
Microscope slide storage box
Microscope slide storage box
Catalog # 631-1090
Supplier:  Azpack
undefined
Microscope slide storage box
Catalog # 631-1090
Supplier:  Azpack
Specifications
  • No. of slides:
    108
  • Colour:
    White/black
  • Description:
    Slide storage box
  • Material:
    Cardboard
  • Pk:
    1
Specifications
About this item
Hand made microscope slide storage box with lid and drop down front.
  • Made from cloth banded cardboard
  • Can hold up to twelve wood pulp trays
  • Each inner tray can hold nine standard slides