


4-Bromophenyl isocyanate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2493-02-9
:
Formula:C₇H₄BrNO
MDL Number:MFCD00002022
:198.02 g/mol
Merck Index:14,1430
Boiling Pt:157…158 °C (14 mmHg)
Melting Pt:39…42 °C
Flash Pt:109 °C (228 °F)
EINECS:219-662-9
UN:2811
ADR:6.1,II
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together