2-Bromo-6-fluoroaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65896-11-9
:
Formula:C₆H₅BrFN
MDL Number:MFCD01310982
:190.01 g/mol
Density:1,674
UN:2810
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together