
2-Bromo-5-fluorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:394-28-5
:
Formula:C₇H₄BrFO₂
MDL Number:MFCD00142874
:219.01 g/mol
Melting Pt:154…158 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together