1,2-Dibromotetrafluorobenzene ≥99%
Catalog # A16620.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:307.87 g/mol
Boiling Pt:197…199 °C
Melting Pt:12 °C
Density:2,26
Flash Pt:99 °C (210 °F)
Storage Temperature:Ambient
EINECS:212-564-7
:GHS07
CAS Number:827-08-7
Formula:C₆Br₂F₄
MDL Number:MFCD00039209