
Methyl-2-bromobenzoate 98%
Catalog # ACRO249810500
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5575 to 1.5595 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
About this item
:
Boiling Pt:253 °C (1013 hPa)
Density:1,51 g/cm³
:215.05 g/mol
EINECS:210-241-5
:GHS07
CAS Number:610-94-6
Formula:C₈H₇BrO₂
MDL Number:MFCD00016328