
Ethyl-2-bromobenzoate 98%
Catalog # ACRO308530250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5430 to 1.5450 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
About this item
:
:229.07 g/mol
Boiling Pt:117 °C (17 mmHg)
Density:1,44 g/cm³
Flash Pt:>110 °C
EINECS:228-034-3
:GHS07
Storage Temperature:Ambient
CAS Number:6091-64-1
Formula:C₉H₉BrO₂
MDL Number:MFCD00015443