
3-Methoxyphenylboronic acid
Catalog # MOLE10126175-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:151.96 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:10365-98-7
Formula:C₇H₉BO₃
MDL Number:MFCD00161359